Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD203c, Mouse Anti-Human, RY610, Clone: NP4D6, BD OptiBuild™

Catalog No. BDB759647 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg;
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB759647 50 μg;
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB759647 Supplier BD Biosciences Supplier No. 759647
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD203c Monoclonal antibody specifically detects CD203c in Human (Tested in Development) samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD203c
Applications Flow Cytometry
Classification Monoclonal
Clone NP4D6
Concentration 0.2 mg/mL
Conjugate RY610
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias ENPP3; E-NPP 3; NPP3; PDNP3; PD-Ibeta; PD-1β; B10; NPPase; gp130RB13-6
Host Species Mouse
Immunogen Human CD203c Transfected Cell Line
Purification Method The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography.
Quantity 50 μg;
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5169
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.