Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD209a Monoclonal Antibody (LWC06), eBioscience™, Invitrogen™
GREENER_CHOICE

Catalog No. 5012985
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-129-85 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-129-85 Supplier Invitrogen™ Supplier No. 14209282
  • $467.00
    $350.65 / Each of 1

    Save $116.35

    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rat Monoclonal Antibody

Description: The LWC06 antibody was generated by immunization with the recombit extracellular region of mouse CIRE/DC-SIGN (CD209). CIRE/DC-SIGN was identified by its expression on CD8 alpha- dendritic cells and plasmacytoid predendritic cells, and is the closest homologue of human DC-SIGN. Human DC-SIGN was originally identified in human placenta for its ability to bind the HIV envelope protein gp120 in a CD4-independent manner. CIRE/DC-SIGN is a 33 kDa type II transmembrane C-type lectin protein. It contains a C-terminal, extracellular, Carbohydrate Recognition Domain (CRD) that is predicted to bind mannose and other carbohydrates in a calcium dependent manner. It has been postulated that CIRE/DC-SIGN may play a role in T-dendritic cell interactions through binding with members of the ICAM family. CIRE/DC-SIGN is differentially expressed by sub-populations of dendritic cells and preliminary data suggest that its expression varies depending on the activation state of the host. CIRE/DC-SIGN is down-regulated in spleen-derived dendritic cell cultures supplemented with GM-CSF. While human DC-SIGN is predomitly expressed in dendritic cells, CIRE/DC-SIGN mRNA has also been detected in B cells. The LWC06 monoclonal antibody does not cross-react with the closely related SIGNR1, SIGNR2, SIGNR3 or SIGNR4. CD209 protein is sensitive to collagenase treatment. Applications Reported: This LWC06 antibody has been reported for use in immunoprecipitation, and immunoblotting (WB).

This gene encodes a transmembrane receptor and is often referred to as DC-SIGN because of its expression on the surface of dendritic cells and macrophages. The encoded protein is involved in the innate immune system and recognizes numerous evolutionarily divergent pathogens ranging from parasites to viruses with a large impact on public health. The protein is organized into three distinct domains: an N-terminal transmembrane domain, a tandem-repeat neck domain and C-type lectin carbohydrate recognition domain. The extracellular region consisting of the C-type lectin and neck domains has a dual function as a pathogen recognition receptor and a cell adhesion receptor by binding carbohydrate ligands on the surface of microbes and endogenous cells. The neck region is important for homo-oligomerization which allows the receptor to bind multivalent ligands with high avidity. Variations in the number of 23 amino acid repeats in the neck domain of this protein are rare but have a significant impact on ligand binding ability. This gene is closely related in terms of both sequence and function to a neighboring gene (GeneID 10332; often referred to as L-SIGN). DC-SIGN and L-SIGN differ in their ligand-binding properties and distribution. Alternative splicing results in multiple variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD209a
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone LWC06
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene Cd209a
Gene Accession No. Q91ZX1
Gene Alias CD209; CD209 antigen; CD209 antigen-like protein A; CD209 molecule; Cd209a; CD209a antigen; CD209a molecule; Cd209d; CD209d antigen; CDSIGN; Cire; CLEC4L; Clec4m; C-type lectin domain family 4 member L; C-type lectin domain family 4, member L; C-type lectin domain family 4, member M; Dcsign; DC-SIGN; DC-SIGN1; Dc-signr; DC-SIGN-related protein; Dendritic cell-specific ICAM-3-grabbing non-integrin; dendritic cell-specific ICAM-3-grabbing non-integrin 1; dendritic cell-specific intercellular adhesion molecule-3-grabbing non-integrin; dendritic cell-specific intracellular adhesion molecules (ICAM)-3 grabbing non-integrin; HIV gpl20-binding protein; MGC129965; MGC130443; RGD1561104; SIGN-R1; SIGNR5
Gene Symbols Cd209a
Host Species Rat
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 170786
Target Species Mouse
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.