Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD209b (SIGN-R1) Monoclonal Antibody (eBio22D1 (22D1)), eBioscience™
GREENER_CHOICE

Catalog No. 5012987
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-129-87 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-129-87 Supplier Invitrogen™ Supplier No. 14209382
Add to Cart
Edge
Add to Cart

Armenian Hamster Monoclonal Antibody

Description: The eBio22D1 monoclonal antibody reacts with mouse SIGNR1 (CD209b). SIGNR1 is a type II transmembrane C-type lectin that was identified in a search for mouse homologues of human DC-SIGN. It is expressed at high levels in splenic marginal zone macrophages and lymph node medullary macrophages, where it functions to uptake dextran polysaccharides, including the capsular polysaccharide of Streptococcus pneumoniae. It has also been demonstrated that SIGNR1 physically associates with TLR4/MD2, and it has been suggested that this association plays a role in recognition of LPS. Furthermore, recently it has been shown that SIGNR1 deficient mice have a defect in catabolism of the complement component C3, and that SIGNR1 binds directly to the complement C1 subcomponent, C1q to assemble a non-conventional C3 convertase. The eBio22D1 monoclonal antibody does not cross-react with the closely related SIGNR1, SIGNR2, SIGNR3 or SIGNR4. Applications Reported: This eBio22D1 (22D1) antibody has been reported for use in flow cytometric analysis, immunoprecipitation, immunohistology staining of frozen tissue sections, and ELISA. (Please use Functional Grade purified eBio22D1 (22D1), Product # 16-2093, in functional assays). Applications Tested: This eBio22D1 (22D1) antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 1 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in...

SIGNR1 is a type II transmembrane C-type lectin that was identified in a search for mouse homologues of human DC-SIGN. It is expressed at high levels in splenic marginal zone macrophages and lymph node medullary macrophages, where it functions to uptake dextran polysaccharides, including the capsular polysaccharide of Streptococcus pneumoniae. It has also been demonstrated that SIGNR1 physically associates with TLR4/MD2, and it has been suggested that this association plays a role in recognition of LPS. Furthermore, recently it has been shown that SIGNR1 deficient mice have a defect in catabolism of the complement component C3, and that SIGNR1 binds directly to the complement C1 subcomponent, C1q to assemble a non-conventional C3 convertase.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD209b (SIGN-R1)
Applications ELISA, Flow Cytometry, Functional Assay, Immunohistochemistry (Frozen), Immunoprecipitation
Classification Monoclonal
Clone eBio22D1 (22D1)
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene Cd209a
Gene Accession No. Q8CJ91, Q91ZX1
Gene Alias 1810030I22Rik; CD209; CD209 antigen-like protein A; CD209 antigen-like protein B; Cd209a; CD209a antigen; CD209a molecule; Cd209b; CD209b antigen; CDSIGN; CIRE; DC SIGNR1; Dcsign; DC-SIGN; DC-SIGN1; DC-SIGNR1; DC-SIGN-related protein 1; Dendritic cell-specific ICAM-3-grabbing non-integrin; MGC130443; mSIGNR1; OtB7; SIGNR1; SIGN-R1; SIGNR5
Gene Symbols Cd209a, Cd209b
Host Species Armenian Hamster
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 170786, 69165
Target Species Mouse
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.