Learn More
Diagnostic Biosystems CD21 CD21
Supplier:  Diagnostic Biosystems PDM122
Encompass_Preferred
This antibody reacts with CD21 antigen of MW 145 kDa, present on the surface of mature B cells. The antigen is present at high density on follicular dendritic cells. CD21 is involved in transmitting growth-promoting signals to the interior of the B cell.
Catalog No. 50-238-5815
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.