Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD217 (IL-17Ra) Monoclonal Antibody (PAJ-17R), APC, eBioscience™, Invitrogen™
GREENER_CHOICE

Catalog No. 5015314
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-153-14 100 μg
50-153-13 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-153-14 Supplier Invitrogen™ Supplier No. 17718282
  • $483.00
    $362.65 / Each of 1

    Save $120.35

    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rat Monoclonal Antibody

Description: The monoclonal antibody PAJ-17R recognizes mouse IL-17 receptor A (IL-17RA) also known as CD217. This single transmembrane domain-containing receptor can homo- or heterodimerize with IL-17RC or IL-17RB. IL-17RA binds to IL-17A, IL-17F(cytokines of the Th 17 lineage), or IL-17E (IL-25). Ligand binding to the single subunit of IL-17RA has been shown to induce homo- or heterodimerization of the receptor complex that in turn recruits Act-1 via the SEFIR domains leading to recruitment of TRAF6 and signaling through the NFkappaB and MAPK pathways. Initial reports suggested ubiquitous expression of IL-17RA based on binding studies with IL-17-Fc and expression of mRNA. Interestingly, despite this, not all cells expressing the receptor respond to IL-17A. In particular, resting T cells do not respond. It is not known if the expression level of IL-17RA protein varies or if some cells express the receptor but are not able to activate downstream signaling events. Recent evidence suggests a more limited expression profile based on IL-17RA knock-out mice which display decreased numbers of neutrophils with impaired function in response to challenge with K. pneumonia or Candida albicans. This suggests IL-17RA plays a more domit and critical role in neutrophil development and function. Our studies suggest that expression can be found on abundantly on Gr-1^dim population and many of the Gr-1^bright bone marrow cells, as well as some F4/80 positive cells.

Interleukin 17A (IL-17A) is a proinflammatory cytokine secreted by activated T-lymphocytes. It is a potent inducer of the maturation of CD34-positive hematopoietic precursors into neutrophils. IL-17RA is a ubiquitous type I membrane glycoprotein that binds with low affinity to IL-17A and IL-17F. IL-17A and its receptor play a pathogenic role in many inflammatory and autoimmune diseases such as rheumatoid arthritis. Defects in IL-17RA are the cause of familial candidiasis type 5 (CANDF5) which is a rare disorder with altered immune responses and impaired clearance of fungal infections.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD217 (IL-17Ra)
Applications Flow Cytometry
Classification Monoclonal
Clone PAJ-17R
Concentration 0.2 mg/mL
Conjugate APC
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene Il17ra
Gene Accession No. Q60943
Gene Alias AW538159; CANDF5; CD127; CD217; CDw217; hIL-17R; IL-17 receptor A; IL17R; Il17ra; IL-17RA; interleukin 17 receptor A; interleukin-17 receptor A; MGC10262; VDw217
Gene Symbols Il17ra
Host Species Rat
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16172
Target Species Mouse
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.