Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD218a (IL-18Ra) Monoclonal Antibody (H44), APC, eBioscience™
GREENER_CHOICE

Catalog No. 501123187
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-112-3187 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-112-3187 Supplier Invitrogen™ Supplier No. 17718342
Add to Cart
Edge
Add to Cart

Mouse Monoclonal Antibody

Description: The H44 antibody reacts with the interleukin-18 receptor alpha chain, also known as IL-1Rrp, an IL-1R family member. IL-18R alpha is expressed on Th1 cells, NK cell subset, neutrophils and IL-12-stimulated tonsillar B cells. By itself, IL-18R alpha binds IL-18 with low affinity and when complexed with IL-18R beta chain binds IL-18 with high affinity. The proinflammatory cytokine IL-18 and its receptor complex play an important role in the innate and autoimmune responses. Applications Reported: This H44 antibody has been reported for use in flow cytometric analysis. Applications Tested: This H44 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (0.25 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended to stain whole blood before RBC lysis. Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser. Filtration: 0.2 μm post-manufacturing filtered.

IL18R1 is a cytokine receptor that belongs to the interleukin 1 receptor family. This receptor specifically binds interleukin 18 (IL18), and is essential for IL18 mediated signal transduction. IFN-alpha and IL12 are reported to induce the expression of this receptor in NK and T cells. The gene along with four other members of the interleukin 1 receptor family, including IL1R2, IL1R1, ILRL2 (IL-1Rrp2), and IL1RL1 (T1/ST2), form a gene cluster on chromosome 2q.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD218a (IL-18Ra)
Applications Flow Cytometry
Classification Monoclonal
Clone H44
Concentration 5 μL/Test
Conjugate APC
Formulation PBS with BSA and 0.09% sodium azide; pH 7.2
Gene Il18r1
Gene Accession No. Q13478
Gene Alias CD218 antigen-like family member A; CD218a; CDw218a; cytokine receptor; IL1 receptor-related protein; Il18r1; IL-18R1; IL-18R-1; IL18RA; Il18ralpha; IL-18R-alpha; IL18Ralpha2; IL1RRP; IL-1Rrp; IL1R-rp; interleukin 1 receptor related protein; interleukin 18 receptor 1; Interleukin-18; interleukin-18 receptor 1; Interleukin-18 receptor alpha
Gene Symbols Il18r1
Host Species Mouse
Purification Method Affinity chromatography
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8809
Target Species Human
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.