Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD218a (IL-18Ra) Monoclonal Antibody (P3TUNYA), APC, eBioscience™, Invitrogen™
GREENER_CHOICE

Catalog No. 17518382
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
17-518-382 100 μg
17-518-380 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 17-518-382 Supplier Invitrogen™ Supplier No. 17518382
Add to Cart
Edge
Add to Cart

Rat Monoclonal Antibody

Description: This P3TUNYA monoclonal antibody reacts with mouse CD218a (Interleukin 18 Receptor alpha). CD218a is one of the subunits of the heterodimeric receptor complex for the IL-18 cytokine. IL-18 is a proinflammatory cytokine and member of the IL-1 family first identified by its role in IFN gamma secretion during toxic shock. Like the other members of the family, it is synthesized as an inactive precursor which requires cleavage by caspases prior to secretion of the active form. It is secreted by activated monocytes, macrophages, epithelial cells, and keratinocytes via a non-classical pathway. IL-18 Receptor is expressed on Th1, but not naive CD4 T cells or Th2, as well as subsets of NK cells and neutrophils. The heterodimeric receptor is made up of a ligand-binding alpha chain and signal-transducing beta chain. On its own, IL-18Ra binds IL-18 with low affinity, although the affinity of this interaction increases upon forming a complex with IL-18Rb. IL-18R signaling plays a critical role in Th1 differentiation and effector function, and results in the increased expression of IFN gamma, TNF alpha, and Fas ligand from T cells and NK cells. IL-18 has also been found to synergize with IL-23 in the development and function of Th17 cells. Recently, a subset of CD8 T cells has been discovered that co-secretes IFN gamma with IL-17A and sometimes IL-22. These Tc17 cells share key transcription factors, chemokine receptors, and cytokine receptors with Th17 cells.

IL18R1 is a cytokine receptor that belongs to the interleukin 1 receptor family. This receptor specifically binds interleukin 18 (IL18), and is essential for IL18 mediated signal transduction. IFN-alpha and IL12 are reported to induce the expression of this receptor in NK and T cells. The gene along with four other members of the interleukin 1 receptor family, including IL1R2, IL1R1, ILRL2 (IL-1Rrp2), and IL1RL1 (T1/ST2), form a gene cluster on chromosome 2q.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD218a (IL-18Ra)
Applications Flow Cytometry
Classification Monoclonal
Clone P3TUNYA
Concentration 0.2 mg/mL
Conjugate APC
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene Il18r1
Gene Accession No. Q61098
Gene Alias CD218 antigen-like family member A; CD218a; CDw218a; cytokine receptor; IL1 receptor-related protein; Il18r1; IL-18R1; IL-18R-1; IL18RA; Il18ralpha; IL-18R-alpha; IL18Ralpha2; IL1RRP; IL-1Rrp; IL1R-rp; interleukin 1 receptor related protein; interleukin 18 receptor 1; Interleukin-18; interleukin-18 receptor 1; Interleukin-18 receptor alpha
Gene Symbols Il18r1
Host Species Rat
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16182
Target Species Mouse
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.