Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD22 Mouse anti-Human, FITC, Clone: HIB22, BD

Catalog No. p-7092480 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB561771 25 Tests
BDB555424 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB561771 Supplier BD Biosciences Supplier No. 561771
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The HIB22 monoclonal antibody specifically binds to CD22. CD22 is a 130-140 kDa glycosylated type I integral membrane protein present on the surface of mature B cells. CD22 is expressed in the cytoplasm of virtually all B cells except plasma cells. CD45RO antigen on T cells and CD75 antigen on B cells have been identified as ligands for CD22. CD22 has been reported to participate in B-cell activation and also as an adhesion molecule. Although the immunobiology of this antigen has not been fully elucidated, reports indicate that ligation of CD22 induces constitutive internalization of the molecule followed by complete degradation.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD22
Applications Flow Cytometry
Classification Monoclonal
Clone HIB22
Conjugate FITC
Description BL-CAM; Siglec-2; Bgp135; Lyb8; LPAP
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Purification Method Affinity Purified
Quantity 25 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.