Learn More
Enzo Life Sciences CD22 Recombinant monoclonal antibody (hL22) (Human IgG1 kappa) (200µg)
SDP
Supplier:  Enzo Life Sciences ENZABS6190200
Encompass_Preferred
Clone: hL22. Isotype: IgG1κ. Immunogen: This antibody was prepared by the humanization of LL2 (EPB-2), a murine anti-CD22 IgG2a raised against Raji Burkitt lymphoma cells. Murine sequences comprise 5–10% of the molecule, with the remainder being human framework sequences, which greatly reduces the potential for immunogenicity. Species reactivity: Human Rhesus monkey, Cynomolgus. Specificity: Recognizes the 3rd Ig-like domain of human CD22 (epitope B), a cell surface glycoprotein present on mature B-cells and on many types of malignant B-cells. Applications: Flow Cytometry, IP, FUNC. Purity: Protein A affinity purified. Formulation: Liquid. In PBS containing 0.02% Proclin 300.
Catalog No. 50-201-3143
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.