Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD226 (DNAM-1) Monoclonal Antibody (10E5), APC, eBioscience™

Catalog No. 17226180
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
17-226-180 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-226-180 Supplier Invitrogen™ Supplier No. 17226180
Add to Cart
Add to Cart

Rat Monoclonal Antibody

This 10E5 monoclonal antibody reacts with mouse CD226, which is also known as DNAX accessory molecule-1 (DNAM-1). CD226 is an adhesion molecule expressed primarily on T cells, including thymocytes. The highest levels of CD226 are found on naive CD8 T cells. Moreover, expression of CD226 is regulated differentially in CD4 T cells. Under certain culture conditions, CD226 can be upregulated in Th1 and downregulated in Th2 and Th0 cells. This cell surface receptor can also be detected on natural killer cells, some B cell subsets, monocytes, and platelets. Interaction with its ligands, the poliovirus receptor (CD155) and CD112, results in adhesion and cell signaling events that promote innate and adaptive immunity, including differentiation, survival, and cytotoxicity. The 10E5 monoclonal antibody has been reported to inhibit antigen-specific T cell proliferation and IFN-gamma production in vivo. Applications Reported: This 10E5 antibody has been reported for use in flow cytometric analysis. Applications Tested: This 10E5 antibody has been tested by flow cytometric analysis of stimulated mouse splenocytes. This can be used at less than or equal to 0.25 μg per test. A test is defined as the amount ( μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the a...

DNAM 1 is a receptor expressed by peripheral blood T lymphocytes that is involved in intercellular adhesion, lymphocyte signaling, cytotoxicity and lymphokine secretion mediated by cytotoxic T lymphocytes. DNAM 1 is broadly expressed on T cells, NK cells, platelets, monocytes and a subset of B cells. DNAM-1 is also expressed by a subset of CD3 positive thymocytes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD226 (DNAM-1)
Applications Flow Cytometry
Classification Monoclonal
Clone 10E5
Concentration 0.2 mg/mL
Conjugate APC
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene Cd226
Gene Accession No. Q8K4F0
Gene Alias adhesion glycoprotein; BC051526; Cd226; CD226 antigen; CD226 molecule; DNAM 1; DNAM1; DNAM-1; DNAX accessory molecule 1; DNAX accessory molecule-1; platelet and T cell activation antigen 1; platelet and T-cell activation antigen 1; Pta1; T lineage-specific activation antigen 1 antigen; TLiSA1
Gene Symbols Cd226
Host Species Rat
Purification Method Affinity chromatography
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 225825
Target Species Mouse
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG2b κ
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.