Learn More
Invitrogen™ CD23 Monoclonal Antibody (LVNC-4)

Description
CD23 Monoclonal Antibody for Flow
Specifications
Specifications
| Antigen | CD23 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | LVNC-4 |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.08% sodium azide |
| Gene | FCER2 |
| Gene Accession No. | P06734 |
| Gene Alias | BLAST-2; CD23; CD23 antigen; CD23A; CLEC4J; C-type lectin domain family 4 member J; C-type lectin domain family 4, member J; Fc epsilon receptor II; FC epsilon RII; Fc fragment of IgE receptor II; Fc fragment of IgE, low affinity II, receptor for (CD23); Fc receptor alpha multiple ligand receptor; Fc receptor, IgE, low affinity II, alpha polypeptide; Fcalpha muR; Fcalpha RII; Fce2; fc-epsilon-RII; FCER2; Fcer2a; FceRII; IGEBF; Immunoglobulin E-binding factor; immunoglobulin epsilon-chain; low affinity immunoglobulin epsilon Fc receptor; Low affinity immunoglobulin epsilon Fc receptor membrane-bound form; Low affinity immunoglobulin epsilon Fc receptor soluble form; low-affinity IgE receptor; Ly-42; lymphocyte IgE receptor |
| Gene Symbols | FCER2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.