Learn More
Invitrogen™ CD243 (ABCB1) Monoclonal Antibody (UIC2), PE, eBioscience™

Description
Description: This monoclonal antibody reacts with human Multidrug Resistant (MDR)-1, which is also known as P-glycoprotein (Pgp) and CD243. A 170-kDa transmembrane protein, MDR-1 is an ATP-dependent efflux pump for lipophilic compounds, including anti-cancer drugs. Expression of MDR-1 has been shown to correlate with multidrug resistance. In fact, tumor resistance to chemotherapy has been linked to MDR-1 expression in many cancers. MDR-1 is expressed in a variety of tissues, including the brain, kidney, liver, pancreas, and testes. Within the immune system, this molecule can be found on normal T, B, and natural killer cells, but not on monocytes. This antibody has been reported to inhibit MDR1-mediated efflux. Applications Reported: This UIC2 antibody has been reported for use in flow cytometric analysis. Applications Tested: This UIC2 antibody has been pre-titrated and tested by flow cytometric analysis on multidrug resistant cell lines. This can be used at 5 μL (0.5 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser. Filtration: 0.2 μm post-manufacturing filtered.
Specifications
Specifications
| Antigen | CD243 (ABCB1) |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | UIC2 |
| Concentration | 5 μL/Test |
| Conjugate | PE |
| Formulation | PBS with BSA and 0.09% sodium azide; pH 7.2 |
| Gene | ABCB1 |
| Gene Accession No. | P08183 |
| Gene Alias | ABC superfamily; P-glycoprotein; ABC20; ABCB1; Abcb1a; Abcb1b; Abcb4; ATP binding cassette subfamily B member 1; ATP-binding cassette sub-family B (MDR/TAP) member 1 (P-glycoprotein/multidrug resistance 1); ATP-binding cassette sub-family B member 1; ATP-binding cassette sub-family B member 1A; ATP-binding cassette sub-family B member 1B; ATP-binding cassette transporter subfamily B, member 1; ATP-binding cassette, sub-family B (MDR/TAP), member 1; ATP-binding cassette, sub-family B (MDR/TAP), member 1 (P-glycoprotein/multidrug resistance 1); ATP-binding cassette, subfamily B (MDR/TAP), member 1A; ATP-binding cassette, sub-family B (MDR/TAP), member 1A; ATP-binding cassette, subfamily B (MDR/TAP), member 1B; ATP-binding cassette, sub-family B (MDR/TAP), member 1B; ATP-binding cassette, sub-family B (MDR/TAP), member 4; ATP-binding cassette, subfamily B, member 1A; ATP-binding cassette, subfamily B, member 1B; ATP-dependent translocase ABCB1; ATP-dependent translocase ABCB1; multidrug resistance protein 1; bovine p-glycoprotein; CD243; CLCS; colchicin sensitivity; doxorubicin resistance; ecotropic viral integration site 32; Evi32; GP170; mdr; Mdr1; MDR1A; Mdr1b; Mdr3; mdr-3; multi-drug resistance 3; multidrug resistance p-glycoprotein; multidrug resistance protein 1; Multidrug resistance protein 1A; multidrug resistance protein 1B; Multidrug resistance protein 2; multidrug resistance protein 3; multidrug resistance protein MDR1; multiple drug resistant 1a; P glycoprotein 1; P glycoprotein 3; P-glycoprotein; P-glycoprotein 1; P-glycoprotein 2; P-glycoprotein 3; P-glycoprotein/multidrug resistance 1; Pgp; p-gp; PGP1; PGP2; PGY1; Pgy-1; Pgy1-1; PGY2; Pgy3; Pgy-3; Phospholipid transporter ABCB1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.