Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD248 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA520436
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA520436 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA520436 Supplier Invitrogen™ Supplier No. PA520436
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

A suggested positive control is human colon tissue lysate. PA5-20436 can be used with blocking peptide PEP-0553. The PA5-20436 immunogen is located within the last 50 amino acids of TEM-1. Predicted molecular ~ 81kD and 46kD. In Western blot applications, this antibody has been observed to detect a band at: 90 kD (post-modifications: 27 O-linked glycosylations) and 46kD. Predicted species reactivity based on immunogen sequence: Mouse (86%); Rat: (86%).

CD248 (CD248 Molecule) is a Protein Coding gene. Gene Ontology (GO) annotations related to this gene include calcium ion binding and extracellular matrix binding. An important paralog of this gene is CD93. CD248 may play a role in tumor angiogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD248
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene CD248
Gene Accession No. Q9HCU0
Gene Alias 2610111G01Rik; AI842296; CD164 sialomucin-like 1; CD164L1; Cd248; CD248 antigen, endosialin; CD248 molecule; CD248 molecule, endosialin; endosialin; MGC119478; MGC119479; TEM1; tumor endothelial marker 1
Gene Symbols CD248
Host Species Rabbit
Immunogen A 14 amino acid peptide near the carboxy terminus of the human TEM1.
Purification Method Antigen Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57124
Target Species Human
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.