Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD25 Monoclonal Antibody (PC61.5), Brilliant Violet™ 711, eBioscience™, Invitrogen™

Catalog No. p-7241378
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
40-702-5180 25 μg
40-702-5182 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 40-702-5180 Supplier Invitrogen™ Supplier No. 407025180
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

CD25 (IL2 receptor alpha chain/IL2RA) is a cytokine that plays a role in the proliferation of T and B lymphocytes. The receptor of this cytokine (IL2RA) is a heterotrimeric protein complex with a gamma chain also shared by interleukin 4 (IL4) and interleukin 7 (IL7). IL2RA, IL2R beta chain (IL2RB), and the IL2R gamma chain (IL2RG), constitute the high-affinity IL2 receptor. Homodimeric IL2RA chains result in low-affinity receptor, while homodimeric IL2RB chains produce a medium-affinity receptor. The expression of IL2 in mature thymocytes is monoallelic, which represents an unusual regulatory mode for controlling the precise expression of a single gene. IL2 is primarily produced by mature T cells. IL2 plays an important role as a growth factor, differentiation factor, and regulator of cell death. IL-2 stimulates the proliferation of B cells, augments natural killer cell activity, and inhibits granulocyte macrophage colony formation. The targeted disruption of a similar gene in mice leads to ulcerative colitis-like disease, which suggests a role in the immune response to antigenic stimuli. Mutations in this gene are associated with interleukin 2 receptor alpha deficiency.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD25
Applications Flow Cytometry
Classification Monoclonal
Clone PC61.5
Concentration 0.2 mg/mL
Conjugate Brilliant Violet 711
Formulation PBS with BSA and 0.09% sodium azide; pH 7.2
Gene IL2RA
Gene Accession No. P01590
Gene Alias ADA; ADA1; Adenosine aminohydrolase; adenosine deaminase; CD25; CD25 antigen; cytokine receptor; IDDM10; IL 2 RA; IL 2 receptor; IL 2R; IL 2R subunit alpha; IL2 RA; IL2 RA antibody; IL2 receptor; il-2 receptor alpha; IL-2 Receptor alpha chain; IL-2 receptor alpha subunit; IL2 receptor subunit alpha; IL-2 receptor subunit alpha; IL2R; IL-2R alpha; IL-2R alpha chain; IL2R subunit alpha; IL-2R subunit alpha; Il2ra; IL-2RA; IL-2-RA; IL2-RA; Il2ra antibody; IL2RAC; IMD41; interleukin 2 receptor; interleukin 2 receptor alpha chain; interleukin 2 receptor subunit alpha; interleukin 2 receptor, alpha; interleukin 2 receptor, alpha chain; interleukin-2 receptor alpha; interleukin-2 receptor alpha chain; interleukin-2 receptor alpha subunit; interleukin-2 receptor beta chain; Interleukin2 receptor subunit alpha; interleukin-2 receptor subunit alpha; Ly-43; p55; p55 chain; sIL 2R; soluble IL 2 receptor; TAC antigen; TCGFR
Gene Symbols IL2RA
Host Species Rat
Purification Method Affinity chromatography
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16184
Target Species Mouse
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG1 λ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.