Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD257 (BAFF, BLyS) Monoclonal Antibody (1D6), FITC, eBioscience™
GREENER_CHOICE

Catalog No. p-7169516
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-901-742 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-901-742 Supplier Invitrogen™ Supplier No. 11901742
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Description: The 1D6 monoclonal antibody reacts with human BLyS (B lymphocyte stimulator), also known as BAFF (B cell activating factor from the TNF family), TALL-1 (TNF and apoptosis leukocyte-expressed ligand-1), THANK (TNF homologue that activates apoptosis, nuclear factor KappaB and c-Jun NH2-terminal kinase) and zTNF4. BLyS, a member of the TNF family, is a type II membrane protein that exists in both membrane-bound and soluble forms. Expression of membrane-bound BLyS on monocytes lineage is regulated by IFN-gamma. BLyS acts as a potent B cell growth factor and costimulator of Ig production. BLyS transgenic mice develop severe B cell hyperplasia and autoimmune lupus-like disease characterized by the presence of autoantibodies against nuclear antigens and immune complexes deposited in the kidney. Data from animal models imply a role for BLyS in human autoimmune diseases. To date BCMA, TACI and BR3 have been identified as receptors for BLyS. APRIL, another TNF family member, and BLyS can form active heterotrimeric molecules when coexpressed and these circulating heterotrimers are present in serum samples from patients with systemic immune-based rheumatic diseases. Furthermore, it has been reported that BLyS and APRIL, share BCMA and TACI as their receptors. Applications Reported: The 1D6 antibody has been reported for use in flow cytometric analysis. Applications Tested: This 1D6 antibody has been pre-titrated and tested by flow cytometric analysis of human BLyS transfe...

BAFF is a membrane bound cytokine expressed by B cell lineage cells. The soluble form of the cytokine binds to the receptors TNFRSF13B/TACI, TNFRSF17/BCMA, and TNFRSF13C/BAFFR. BAFF is a potent B cell activator, and important for the proliferation and differentiation of B cells. BAFF is also expressed by T lymphocytes and has been shown to promote T cell activation and survival. Isoforms representing alternatively spliced, and glycosylated variants have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD257 (BAFF, BLyS)
Applications Flow Cytometry
Classification Monoclonal
Clone 1D6
Concentration 5 μL/Test
Conjugate FITC
Formulation PBS with BSA and 0.09% sodium azide; pH 7.2
Gene TNFSF13B
Gene Accession No. Q9Y275
Gene Alias ApoL related ligand TALL-1; b cell-activating factor; B lymphocyte stimulator; BAFF; B-cell-activating factor; B-lymphocyte stimulator; BLYS; CD257; D8Ertd387e; delta BAFF; Delta4 BAFF; dendritic cell-derived TNF-like molecule; DTL; RGD1561519; TALL1; TALL-1; THANK; TNF- and APOL-related leukocyte expressed ligand 1; TNF and ApoL-related leukocyte expressed ligand 1; TNF homolog that activates apoptosis; Tnfsf13b; TNFSF20; TNLG7A; tumor necrosis factor (ligand) superfamily member 13b; tumor necrosis factor (ligand) superfamily, member 13b; tumor necrosis factor (ligand) superfamily, member 20; tumor necrosis factor ligand 7A; tumor necrosis factor ligand superfamily member 13B; Tumor necrosis factor ligand superfamily member 13b, membrane form; Tumor necrosis factor ligand superfamily member 13b, soluble form; tumor necrosis factor superfamily member 13b; tumor necrosis factor-like protein ZTNF4; UNQ401/PRO738; ZTNF4
Gene Symbols TNFSF13B
Host Species Mouse
Purification Method Affinity chromatography
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10673
Target Species Human
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.