Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD258 (LIGHT) Monoclonal Antibody (7-3 (7)), APC, eBioscience™
GREENER_CHOICE

Catalog No. 5015134
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-151-34 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-151-34 Supplier Invitrogen™ Supplier No. 17258942
  • $452.00
    $339.65 / Each of 1

    Save $112.35

    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

Description: This 7-3 (7) monoclonal antibody reacts with human CD258 (also known as LIGHT), a 29-kDa type II transmembrane protein that is a member of the TNF ligand superfamily related to lymphotoxin-β (LT-β). Expressed on activated T cells, particularly CD8+ T cells, LIGHT plays a role in cell survival, inflammation, and tumor eradication. Furthermore, upon binding its ligand, herpesvirus entry mediator (HVEM), LIGHT induces a costimulatory effect on T cells, enhancing T cell proliferation and cytokine production. LIGHT has also been shown to indirectly interact with the LT-β receptor. Applications Reported: This 7-3 (7) antibody has been reported for use in flow cytometric analysis. Applications Tested: This 7-3 (7) antibody has been tested by flow cytometric analysis on PMA- and ionomycin-stimulated normal human peripheral blood cells. This can be used at 5 μL (0.25 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser. Filtration: 0.2 μm post-manufacturing filtered.

Apoptosis, or programmed cell death, occurs during normal cellular differentiation and development of multicellular organisms. Apoptosis is induced by certain cytokines including TNF (tumor necrosis factor) and Fas ligand of the TNF family through their death domain containing receptors, TNFR1 (tumor necrosis factor receptor 1) and Fas. Cell death signals are transduced by death domain (DD)-containing adapter molecules and members of the ICE (interleukin-1-beta converting enzyme)/CED-3 (C. elegans death receptor-3) protease family. A novel DD-containing molecule was recently cloned from mouse, human and monkey and designated Daxx (death associated protein 6). Daxx binds specifically to the Fas death domain and enhances Fas induced apoptosis and activates the Jun N-terminal kinase (JNK) pathway. Daxx is widely expressed in fetal and adult human and mouse tissues indicating its important function in Fas signaling pathways.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD258 (LIGHT)
Applications Flow Cytometry
Classification Monoclonal
Clone 7-3 (7)
Concentration 5 μL/Test
Conjugate APC
Formulation PBS with BSA and 0.09% sodium azide; pH 7.2
Gene TNFSF14
Gene Accession No. O43557
Gene Alias CD258; Herpes virus entry mediator ligand; herpesvirus entry mediator ligand; HVEML; HVEM-L; Light; LTg; Ly113; sCD258; sLIGHT; soluble CD258; soluble LIGHT; Tnfsf14; TR2; tumor necrosis factor (ligand) superfamily, member 14; tumor necrosis factor ligand 1D; tumor necrosis factor ligand superfamily member 14; Tumor necrosis factor ligand superfamily member 14, membrane form; Tumor necrosis factor ligand superfamily member 14, soluble form; tumor necrosis factor superfamily member 14; UNQ391/PRO726
Gene Symbols TNFSF14
Host Species Mouse
Purification Method Affinity chromatography
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8740
Target Species Human
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.