Learn More
Invitrogen™ CD263 (TRAIL-R3) Monoclonal Antibody (DJR3), PE, eBioscience™

Description
Description: The DJR3 monoclonal antibody reacts with human Decoy receptor 1 (DcR1), a glycophospholipid-anchored cell surface protein also known as TRAIL-R3, TRID, and LIT. DcR1 has an extracellular TRAIL binding domain and lacks the cytoplasmic death domain (DD) shared by several members of the TNF receptor superfamily. DcR1, along with DcR2, protects cells from TRAIL-mediated cytotoxicity. DcR1 binds to TRAIL and inhibits binding of TRAIL to DR4 or DR5. DcR1 is expressed broadly in many normal tissues at low level, but not in most tumors. Overexpression of DcR1 attenuates TRAIL-induced apoptosis. Applications Reported: The DJR3 antibody has been reported for use in flow cytometric analysis. Applications Tested: This DJR3 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells and human DcR1-transfected cells. This can be used at 5 μL (1 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser. Filtration: 0.2 μm post-manufacturing filtered.
Specifications
Specifications
| Antigen | CD263 (TRAIL-R3) |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | DJR3 |
| Concentration | 5 μL/Test |
| Conjugate | PE |
| Formulation | PBS with BSA and 0.09% sodium azide; pH 7.2 |
| Gene | TNFRSF10C |
| Gene Accession No. | O14798 |
| Gene Alias | Antagonist decoy receptor for TRAIL/Apo-2L; CD263; cytotoxic TRAIL receptor-3; DCR1; DCR1-TNFR; Decoy receptor 1; decoy TRAIL receptor without death domain; LIT; lymphocyte inhibitor of TRAIL; MGC149501; MGC149502; TNF receptor superfamily member 10c; TNF-related apoptosis-inducing ligand receptor 3; TNFRSF10C; TRAIL receptor 3; TRAIL receptor without an intracellular domain; TRAILR3; TRAIL-R3; TRID; Tumor necrosis factor receptor superfamily member 10C; tumor necrosis factor receptor superfamily, member 10c, decoy without an intracellular domain; UNQ321/PRO366 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.