Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD266 Recombinant Rabbit Monoclonal Antibody (7H8)
GREENER_CHOICE

Catalog No. PIMA538356
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA538356 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA538356 Supplier Invitrogen™ Supplier No. MA538356
Only null left
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

This antibody has been tested in direct-ELISA.

TWEAK is a member of the TNF family and was first described as a weak inducer of apoptosis in demonstrate that this relatively small molecule is a fully transformed cell lines. Recently, a receptor for TWEAK was isolated by expression cloning from a HUVEC cell cDNA library. TweakR is a small protein belonging to TNF receptor super family. TweakR is capable of intiating a proliferative signal by binding to TRAF1, 2, and 3. TweakR mRNA is upregulated during tissue regeneration in the aorta and liver. It may also play a role in vascular cell migration and angiogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD266
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone 7H8
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 150mM NaCl and 0.02% sodium azide; pH 7.4
Gene TNFRSF12A
Gene Accession No. Q9NP84
Gene Alias AI255180; C87282; CD266; FGF-inducible 14; Fgfrp2; fibroblast growth factor regulated protein 2; fibroblast growth factor-inducible immediate-early response protein 14; Fibroblast growth factor-regulated protein 2; FN14; HPIP; TNF receptor superfamily member 12A; TNFRSF12A; Tumor necrosis factor receptor superfamily member 12A; tumor necrosis factor receptor superfamily, member 12A; TweakR; TWEAK-R; Tweak-receptor; type I transmembrane protein Fn14
Gene Symbols TNFRSF12A
Host Species Rabbit
Immunogen A synthesized peptide derived from human TWEAKR.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51330
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.