Learn More
Invitrogen™ CD267 (TACI) Polyclonal Antibody

Description
Suggested positive control: human spleen, Daudi, or RAW, antigen standard for TNFRSF13B (transient overexpression lysate).
Specifications
Specifications
| Antigen | CD267 (TACI) |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.05% sodium azide |
| Gene | TNFRSF13B |
| Gene Accession No. | O14836, Q9ET35 |
| Gene Alias | 1200009E08Rik; CD267; CVID; CVID2; IGAD2; NEWGENE_1595998; RYZN; TACI; TNF receptor superfamily member 13B; Tnfrsf13b; TNFRSF14B; transmembrane activator and calcium-modulating and cyclophilin ligand interactor; transmembrane activator and CAML interactor; tumor necrosis factor receptor 13B; tumor necrosis factor receptor superfamily member 13B; tumor necrosis factor receptor superfamily, member 13B |
| Gene Symbols | TNFRSF13B |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.