Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD267 (TACI) Rat anti-Mouse, Brilliant Violet 421, Clone: 8F10, BD Optibuild™

Catalog No. BDB742840 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB742840 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB742840 Supplier BD Biosciences Supplier No. 742840
Only null left
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The 8F10 monoclonal antibody specifically recognizes CD267 which is also known as transmembrane activator and calcium-modulating and cyclophilin ligand interactor(transmembrane activator and CAML interactor or TACI). CD267 is encoded by Tnfrsf13b /(tumor necrosis factor receptor superfamily member 13B). CD267 is a type III transmembrane protein receptor that binds to the ligands BAFF (BLyS), and APRIL. BAFF overproduction is associated with lupus erythematosus and Sjφgren's disease, two systemic autoimmune diseases. CD267 is expressed most notably on maturing subsets of splenic B cells such as transition type 2 (T2) and marginal zone (MZ) B cells and also in activated T-cells. In CD267 deficient mice, B cell numbers are increased and mice develop autoimmune disorders suggesting that CD267 plays a role in B cell regulation.

The antibody was conjugated to BD Horizon™ BV421 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 407nm and Em Max at 421nm, BD Horizon BV421 can be excited by the violet laser and detected in the standard Pacific Blue™ filter set (eg, 450/50nm filter). BD Horizon BV421 conjugates are very bright, often exhibiting a 10 fold improvement in brightness compared to Pacific Blue conjugates. BV421 is a polymer-based dye excited by the violet laser and is one of the brightest fluorochromes offered by BD Biosciences. Conjugates are typically 10 times brighter than Pacific Blue™ conjugates and are often as bright as or brighter than PE conjugates. Due to nearly identical excitation and emission properties but different spillover characteristics, BD Horizon BV421, Pacific Blue™, and BD Horizon™ V450 cannot be used simultaneously.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD267 (TACI)
Classification Monoclonal
Clone 8F10
Concentration 0.2mg/mL
Conjugate Brilliant Violet 421
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias TACI, Taci,Tnfrsf13b, transmembrane activator and CAML interactor
Host Species Rat
Immunogen Mouse TACI Extracellular Domain Protein
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.