Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD271 (NGF Receptor) Monoclonal Antibody (ME20.4), Super Bright™ 600, eBioscience™, Invitrogen™
GREENER_CHOICE

Catalog No. p-7159024
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
63-940-042 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 63-940-042 Supplier Invitrogen™ Supplier No. 63940042
Only null left
Add to Cart
Edge
Add to Cart

Mouse Monoclonal Antibody

The monoclonal antibody ME20.4 recognizes CD271, a 75 kDa, type I transmembrane protein that belongs to the low-affinity neurotrophin receptor and tumor necrosis factor receptor superfamily. CD271 is expressed by neurons, Schwann cells, melanocytes, mesen

Neurotrophic factors control the survival, differentiation and maintenance of neurons in the peripheral and central nervous systems, and of other neural crest-derived cell types. Developing sympathetic neurons are absolutely dependent upon nerve growth factor (NGF) during the period of target competition in vivo. Antibodies reacting specifically with NGFR p75 are useful tools in the detection and characterization of NGFR p75, to enhance our understanding of a wide range of phenomena in the development, plasticity and repair of the nervous system.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD271 (NGF Receptor)
Applications Flow Cytometry
Classification Monoclonal
Clone ME20.4
Concentration 5 μL/Test
Conjugate Super Bright 600
Formulation PBS with BSA and 0.09% sodium azide; pH 7.2
Gene Ngfr
Gene Accession No. P07174, P08138, Q9Z0W1
Gene Alias CD271; CD271 antigen; Gp80-LNGFR; LNGFR; low affinity nerve growth factor receptor; low affinity neurotrophin receptor p75NTR; LOW QUALITY PROTEIN: tumor necrosis factor receptor superfamily member 16; low-affinity nerve growth factor receptor; nerve growth factor receptor; nerve growth factor receptor (TNFR superfamily, member 16); nerve growth factor receptor, fast; NGF receptor; NGFR; NGR; p75; p-75; p75 ICD; p75 neurotrophin receptor; p75 ntr; p75(NTR); p75NGFR; p75NTR; RNNGFRR; TNFR superfamily, member 16; Tnfrsf16; TNR16; Tumor necrosis factor receptor superfamily member 16
Gene Symbols Ngfr
Host Species Mouse
Purification Method Affinity chromatography
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100349095, 100525607, 101119064, 18053, 24596, 4804, 491071
Target Species Canine, Human, Mouse, Monkey, Sheep, Pig, Rabbit, Rat
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.