Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD277 Monoclonal Antibody (eBioBT3.1 (20.1, BT3.1)), eBioscience™
GREENER_CHOICE

Catalog No. 5013004
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-130-04 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-130-04 Supplier Invitrogen™ Supplier No. 14277982
  • $317.00
    $237.65 / Each of 1

    Save $79.35

    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

Description: The eBioBT3.1 monoclonal antibody recognizes BT3.1, BT3.2 and BT3.3. The BT family of proteins is a subgroup of the Ig superfamily (IgSF). BT is a glycoprotein that forms a major component of the milk fat globule molecule. In addition to BT, there are six other genes that have been discovered which have been sub-divided into the BT2 (BT2.1, BT2.2, and BT2.3) and BT3 (BT3.1, BT3.2, and BT3.3) families. BT3.1, BT3.2, and BT3.3 share 95% mRNA identity. BT3 molecules are constitutively expressed on the cell surface of antigen-presenting cells, and the IgV-like domain of BT3.1 is sufficient for interaction with its counter-receptor. BT3.1 is expressed by T cells, B cells and CD14+ cells. Applications Reported: This eBioBT3.1 (20.1, BT3.1) antibody has been reported for use in flow cytometric analysis, and immunoprecipitation. Applications Tested: This eBioBT3.1 (20.1, BT3.1) antibody has been tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at less than or equal to 1 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.2 μm post-man...

The butyrophilin (BTN) genes are a group of major histocompatibility complex (MHC)-associated genes that encode type I membrane proteins with 2 extracellular immunoglobulin (Ig) domains and an intracellular B30.2 (PRYSPRY) domain. Three subfamilies of human BTN genes are located in the MHC class I region: the single-copy BTN1A1 gene and the BTN2 and BTN3 genes, which have undergone tandem duplication, resulting in 3 copies of each.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD277
Applications Flow Cytometry, Immunoprecipitation
Classification Monoclonal
Clone eBioBT3.1 (20.1, BT3.1), BT3.1)
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene BTN3A1
Gene Accession No. O00481
Gene Alias BT3.1; BTF5; BTN3.1; BTN3A1; butyrophilin; Butyrophilin subfamily 3 member A1; butyrophilin, subfamily 3, member A1; CD277; dJ45P21.3 (butyrophilin, subfamily 3, member A1)
Gene Symbols BTN3A1
Host Species Mouse
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11119
Target Species Human
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.