Learn More
BD Cell Analysis 3P Co-Stimulatory Antibodies (CD28/CD49d), 0.3 mL
Supplier:  BD Cell Analysis 3P 347690
Encompass
anti-Human CD28/CD49d Purified. Brand: BD FastImmune, Reactivity: Human (QC Testing), application: Flow cytometry (Routinely Tested), Storage Buffer: 0.01% sodium azide. Store at 2 Deg. C to 8 Deg. C.
Catalog No. 50-620-653
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.