Learn More
Sino Biological Human / Cynomolgus / Rhesus CD28 Protein (ECD, His Tag) 50ug
Supplier:  Sino Biological 90182C08H50
Encompass_Preferred
A DNA sequence encoding the human/cynomolgus/rhesus CD28 (P10747-1/Q0PDN3-1/NP_001036106.2) (Asn19-Pro152) was expressed with a polyhistidine tag at the C-terminus and a signal peptide (Q0PDN3-1/NP_001036106.2, Met1-Gly18) at the N-terminus. Human, Cynomolgus and Rhesus CD28 sequences (Asn19-Pro152) are identical.
Catalog No. 50-211-5944
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.