Learn More
Invitrogen™ CD28 Recombinant Rabbit Monoclonal Antibody (007), PerCP

Description
Preparation: This antibody was obtained from a rabbit immunized with purified, recombinant Human CD28 (Met1-Pro152) and conjugated with PerCP under optimum conditions, the unreacted PerCP was removed. Storage: This antibody can be stored at 2C-8C for twelve months without detectable loss of activity. Protected from prolonged exposure to light. Do not freeze!.
Specifications
Specifications
| Antigen | CD28 |
| Applications | Flow Cytometry |
| Classification | Recombinant Monoclonal |
| Clone | 007 |
| Concentration | 100 μg/mL |
| Conjugate | PerCP |
| Formulation | PBS with 0.5% BSA and 0.03% Proclin 300 |
| Gene | Cd28 |
| Gene Accession No. | P10747 |
| Gene Alias | antigen CD28; CD28; CD28 antigen; CD28 antigen (Tp44); CD28 isoform; CD28 isoform 2; Cd28 molecule; CD28 precursor protein; CD28 protein; CD28 protein precursor; CD28RNA; cell surface protein; CHT28; contains partial extracellular domain; costimulatory molecule B7 receptor CD28; EGK_04709; MGC138290; sCD28; soluble CD28; T44; T-cell costimulatory molecule CD28; T-cell costimulatory molecule Tp44; T-cell-specific surface glycoprotein CD28; T-cell-specific surface glycoprotein CD28 homolog; TP44 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.