Learn More
Invitrogen™ CD282 (TLR2) Monoclonal Antibody (TL2.1), Functional Grade, eBioscience™

Description
Description: The TL2.1 monoclonal antibody reacts with human Toll-like receptor 2 (TLR2). To date, at least ten members of the Toll family have been identified in human. This family of type I transmembrane proteins is characterized by an extracellular domain with leucine-rich repeats and a cytoplasmic domain with homology to the type I IL-1 receptor. Two of these receptors, TLR2 and TLR4, are pattern recognition receptors and signaling molecules in response to bacterial lipoproteins and have been implicated in innate immunity and inflammation. TLR2 is expressed by peripheral blood monocytes and is responsible for distinguishing different pathogens. TL2.1, a blocking antibody, has been used to study the role of TLR-2 as a pattern recognition receptor in microbial lipoprotein/lipopeptide induced cytokine production from human peripheral blood mononuclear cells. TL2.1 has been reported to immunoprecipitate human TLR2 (~90 kDa) from PBMC and HMEC. Applications Reported: TL2.1 has been reported for use in flow cytometric analysis and blocking of TLR2-mediated cytokine production. Inhibition of cytokine production has been reported in the literature using Listeria monocytogenes and peptidoglycans. Optimal conditions for the inhibition assays using TL2.1 and specific stimulatory reagents/suppliers have to be evaluated by individual investigators. Neutralization Protocol: (Important note: make sure to eliminate LPS contamination in the culture system) 1) Prepare target cells of in...
Specifications
Specifications
| Antigen | CD282 (TLR2) |
| Applications | Flow Cytometry, Functional Assay, Neutralization |
| Classification | Monoclonal |
| Clone | TL2.1 |
| Concentration | 1 mg/mL |
| Conjugate | Functional Grade |
| Formulation | PBS with no preservative; pH 7.2 |
| Gene | Tlr2 |
| Gene Accession No. | O60603 |
| Gene Alias | CD282; Ly105; TIL4; Tlr2; TLR-2; toll like receptor 2; toll/interleukin-1 receptor-like protein 4; Toll-like receptor; toll-like receptor 2; toll-like receptor 2 variant 1; toll-like receptor 2 variant 2; toll-like-receptor 2, exon 1a_; Toll-like-receptor, exon 1b; Toll-like-receptor, exon 1b_; Toll-like-receptor2, exon 1a |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.