Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD297 Monoclonal Antibody (01)
GREENER_CHOICE

Catalog No. PIMA530093
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA530093 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA530093 Supplier Invitrogen™ Supplier No. MA530093
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Rat CD297.

This gene encodes a protein that contains a mono-ADP-ribosylation motif. It is a member of the ADP-ribosyltransferase gene family but enzymatic activity has not been demonstrated experimentally. Antigens of the Dombrock blood group system are located on the gene product, which is glycosylphosphatidylinosotol-anchored to the erythrocyte membrane. Allelic variants, some of which lead to adverse transfusion reactions, are known.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD297
Applications ELISA
Classification Monoclonal
Clone 01
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene Art4
Gene Accession No. 0
Gene Alias 4432404K01Rik; ADP-ribosyltransferase 4; ADP-ribosyltransferase 4 (DO blood group); ADP-ribosyltransferase 4 (Dombrock blood group); ADP-ribosyltransferase 4 (inactive) (Dombrock blood group); ADP-ribosyltransferase C2 and C3 toxin-like 4; ART4; ARTC4; ATR4; CD297; DO; DOK1; Dombrock blood group carrier molecule; ecto-ADP-ribosyltransferase 4; mono (ADP-ribosyl)transferase; mono(ADP-ribosyl)transferase 4; mono-ADP-ribosyltransferase 4; NAD(+)--protein-arginine ADP-ribosyltransferase 4; NAD(P)(+)--arginine ADP-ribosyltransferase; NAD(P)(+)--arginine ADP-ribosyltransferase 4
Gene Symbols Art4
Host Species Mouse
Immunogen Recombinant Rat CD297 protein.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 312806
Target Species Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.