Learn More
Invitrogen™ CD298 Recombinant Rabbit Monoclonal Antibody (0B7M7)

Description
Positive Samples: 293T, U-87MG, Mouse lung, Mouse testis, Mouse brain, Rat lung, Rat testis, Rat spleen Immunogen sequence: GLKPEGVPRI DCVSKNEDIP NVAVYPHNGM IDLKYFPYYG KKLHVGYLQP LVAVQVSFAP NNTGKEVTVE CKIDGSANLK SQDDRDKFLG RVMFKITARA.
Specifications
Specifications
| Antigen | CD298 |
| Applications | ELISA, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Recombinant Monoclonal |
| Clone | 0B7M7 |
| Concentration | 0.45 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | ATP1B3 |
| Gene Accession No. | P54709, P97370, Q63377 |
| Gene Alias | AA409958; AI664000; Atp1b3; ATPase Na+/K+ transporting subunit beta 3; ATPase, Na+/K+ beta 3 polypeptide; ATPase, Na+/K+ transporting, beta 3 polypeptide; ATPase, Na+K+ transporting, beta polypeptide 3; ATPB-3; AW212096; CD298; CD298 antigen; Na K-ATPase beta-3 subunit; Na+/K+ -ATPase beta 3 subunit; NKAB3S; Sodium Potassium ATPase; Sodium/potassium-dependent ATPase subunit beta-3; sodium/potassium-transporting ATPase subunit beta-3 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.