Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD299 (DC-SIGN/L) Monoclonal Antibody (16E7), APC, eBioscience™
GREENER_CHOICE

Catalog No. 501123118
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-112-3118 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-112-3118 Supplier Invitrogen™ Supplier No. 17299942
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

Description: The 16E7 monoclonal antibody recognizes human CD299, which is also known as DC-SIGNR or L-SIGN. A homolog of DC-SIGN, CD299 is an oligomeric type II transmembrane protein expressed on dendritic cells, macrophages in the lung, and endothelial cells in the liver. This C-type lectin binds ICAM-3 to mediate the interaction between dendritic cells and T cells. CD299 also binds the gp120 protein of HIV and the E2 envelope protein of HCV, thereby playing a role in viral infection. Applications Reported: This 16E7 antibody has been reported for use in flow cytometric analysis. Applications Tested: This 16E7 antibody has been pre-titrated and tested by flow cytometric analysis of normal human monocyte-derived dendritic cells. This can be used at 5 μL (0.125 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser. Filtration: 0.2 μm post-manufacturing filtered.

CLEC4M is a type II integral membrane protein that is 77% identical to CD209 antigen, a HIV gp120-binding protein. This protein, like CD209, efficiently binds both intercellular adhesion molecule 3 (ICAM3) and HIV-1 gp120, and enhances HIV-1 infection of T cells.Western blots using two different antibodies against two unique regions of this protein target confirm the same apparent molecular weight in our tests.This gene encodes a type II integral membrane protein that is 77% identical to CD209 antigen, a HIV gp120-binding protein. This protein, like CD209, efficiently binds both intercellular adhesion molecule 3 (ICAM3) and HIV-1 gp120, and enhances HIV-1 infection of T cells. This gene is mapped to 19p13.3, in a cluster with the CD209 and CD23/FCER2 genes. Multiple alternatively spliced transcript variants have been found for this gene, but the biological validity of some variants has not been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD299 (DC-SIGN/L)
Applications Flow Cytometry
Classification Monoclonal
Clone 16E7
Concentration 5 μL/Test
Conjugate APC
Formulation PBS with BSA and 0.09% sodium azide; pH 7.2
Gene CLEC4M
Gene Accession No. Q9H2X3
Gene Alias CD209 antigen-like protein 1; Cd209b; CD209b antigen; CD209L; CD209L1; CD299; CD299 antigen; CLEC4M; C-type lectin domain family 4 member M; C-type lectin domain family 4, member M; DC-SIGN2; DCSIGNR; DC-SIGNR; DC-SIGN-related protein; Dendritic cell-specific ICAM-3-grabbing non-integrin 2; HP10347; liver/lymph node-specific ICAM-3 grabbing non-integrin; liver/lymph node-specific ICAM-3-grabbing non-integrin; LSIGN; L-SIGN; mannose binding C-type lectin DC-SIGNR; MGC129964; MGC47866
Gene Symbols CLEC4M
Host Species Mouse
Purification Method Affinity chromatography
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10332
Target Species Human
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.