Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD2BP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5117849
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5117849 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5117849 Supplier Invitrogen™ Supplier No. PA5117849
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

CD2BP2 Polyclonal Antibody for Western Blot, IP

This gene encodes a bi-functional protein. In the cytoplasm, the encoded protein binds the cytoplasmic tail of human surface antigen CD2 via its C-terminal GYF domain, and regulate CD2-triggered T lymphocyte activation. In the nucleus, this protein is a component of the U5 small nuclear ribonucleoprotein complex and is involved in RNA splicing. A pseudogene has been identified on chromosome 7. Alternative splicing results in multiple transcript variants but their biological validity has not been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD2BP2
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 0.03% ProClin 300; pH 7
Gene CD2BP2
Gene Accession No. O95400
Gene Alias 1500011B02Rik; 2410024K20Rik; AW048183; CD2 (cytoplasmic tail) binding protein 2; CD2 antigen (cytoplasmic tail) binding protein 2; CD2 antigen cytoplasmic tail-binding protein 2; CD2 cytoplasmic domain-binding protein 2; CD2 cytoplasmic tail binding protein 2; CD2 tail-binding protein 2; CD2BP2; FWP010; KIAA1178; LIN1; PPP1R59; protein phosphatase 1, regulatory subunit 59; Snu40; U5 snRNP 52K protein; U5-52K
Gene Symbols CD2BP2
Host Species Rabbit
Immunogen E. coli-derived Human CD2BP2 fragment.
Purification Method Antigen Affinity Chromatography, Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10421
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.