Learn More
Diagnostic Biosystems CD3 CD3 (F7.2.38)
Supplier:  Diagnostic Biosystems MOB42201
Encompass_Preferred
CD3 is first detectable in early thymocytes and its appearance probably represents one of the earliest signs of commitment to T cell lineage. In cortical thymocytes, the antigen is predominantly present as an intracytoplasmic constituent. This antibody labels CD3ε and is a useful tool for the identification of T cells and related neoplasms.
Catalog No. 50-238-5516
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.