Learn More
Diagnostic Biosystems CD3 CD3 (SP7)
Supplier:  Diagnostic Biosystems RMAB00505
Encompass_Preferred
The CD3 antigen is present on early thymocytes and mature T cells and is generally regarded as a pan-T cell marker. This antibody will help detect CD3 expression in normal and neoplastic tissues. This antibody reacts with the intracytoplasmic portion of the CD3 antigen expressed by T cells. It stains human T cellsin both the cortex and medulla of the thymus and in peripheral lymphoid tissues. This antibody issuitable for staining normal and neoplastic T cells in formalin-fixed, paraffin-embedded tissues.
Catalog No. 50-238-5772
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.