Learn More
Biocare Medical, LLC CD3 [LN10] Antibody - Concentrate, 1.0 ml
SDP
Supplier:  Biocare Medical, LLC ACI3152C
Encompass_Preferred
CD3 is expressed throughout the T-cell differentiation process. CD3 is a highly specific and sensitive T-cell lineage marker, making it ideal for the immunophenotypic analysis of lymphohaematopoietic malignancies. Notable exceptions include some of the more aggressive large T-cell lymphomas and CD30 (Ki-1) positive anaplastic large cell lymphomas, which may not express detectable antigen (1-3). CD3 [LN10] has demonstrated optimal staining when compared to other CD3 clones including PS1, F7.2.38 and SP7 (4). A monoclonal antibody to human CD3 is regarded as a reliable pan T-cell antibody used in the immunophenotyping of lymphomas in paraffin sections.
Catalog No. NC1922809
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.