Learn More
CD3 Mouse anti-Human , BUV615, Clone: UCHT1 (also known as UCHT-1; UCHT 1), BD Biosciences™
Shop All BD Biosciences ProductsDescription
Specifications
Specifications
| Antigen | CD3 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | UCHT1 (also known as UCHT-1; UCHT 1) |
| Conjugate | Brilliant Ultraviolet 615 |
| Formulation | Aqueous buffered solution containing ≤0.09% sodium azide. |
| Gene Alias | CD3e; CD3E; T3E; TCRE; T-cell surface antigen T3/Leu-4 epsilon |
| Host Species | Mouse |
| Immunogen | Human infant thymocytes and peripheral blood lymphocytes from a Sézary Syndrome donor |
| Quantity | 100 Tests |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.