Learn More
Epredia™ Lab Vision™ CD30 (Reed-Sternberg Cell Marker), Mouse Monoclonal Antibody
Shop All Epredia ProductsDescription
Host Species: Mouse
Clone: CON6D/C2
Isotype: IgG2a
Species Reactivity: Human. Others not known.
Immunogen: Hodgkin's lymphoma cell line L428
Molecular Weight: 120kDa (mature), 90kDa (precursor)
Positive Control: Hodgkin's lymphoma
Cellular Localization: Membrane and cytoplasm
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Note:
USA: RUO; Int'l: RUO
Specifications
Specifications
| Antigen | CD30 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | CON6D/ C2 |
| Conjugate | Unconjugated |
| Formulation | Ready-to-use for IHC (UVLP) |
| Host Species | Mouse |
| Immunogen | Hodgkins lymphoma cell line L428 |
| Purification Method | Hodgkin's lymphoma |
| Quantity | 1 mL |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.