Learn More
Invitrogen™ CD300b Polyclonal Antibody

Description
Positive test controls include: Mouse lung, Rat lung. Immunogen sequence: MWLPPALLLL SLSGCFSIQG PESVRAPEQG SLTVQCHYKQ GWETYIKWWC RGVRWDTCKI LIETRGSEQG EKSDRVSIKD NQKDRTFTVT MEGLRRDDAD VYWCGIERRG PDLGTQVKVI VDPEGAASTT ASSPTNSNMA VFIGSHKRNH YMLLVFVKVP ILLILVTAIL WLKGSQRVPE EPGEQPIYMN FSEPLTKDMA T.
Specifications
Specifications
| Antigen | CD300b |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 1.95 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 |
| Gene | CD300LB |
| Gene Accession No. | A8K4G0, Q3U497 |
| Gene Alias | CD300 antigen like family member B; CD300 antigen-like family member B; CD300 molecule like family member b; CD300 molecule-like family member b; CD300b; CD300LB; CLM7; CLM-7; CMRF35A2; CMRF35-A2; CMRF35-like molecule 7; CMRF-35-like molecule-7; Gm253; Immune receptor expressed on myeloid cells 3; IREM3; IREM-3; Leukocyte mono-Ig-like receptor 5; LMIR5; TREM5; TREM-5; triggering receptor expressed on myeloid cells 5; UNQ2530/PRO6029 |
| Gene Symbols | CD300LB |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.