Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD309 (FLK1) Monoclonal Antibody (Avas12a1), Biotin, eBioscience™, Invitrogen™
GREENER_CHOICE

Catalog No. 5011940
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
500 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-119-40 100 μg
50-119-41 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-119-40 Supplier Invitrogen™ Supplier No. 13582182
  • $199.00
    $149.25 / Each of 1

    Save $49.75

    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

Rat Monoclonal Antibody

Description: The Avas12a1 monoclonal antibody reacts with mouse Flk-1, also known as vascular endothelial growth factor receptor 2 (VEGFR2). Flk-1 is a receptor tyrosine kinase involved in vascular endothelial tissue development and is expressed on endothelial cells during embryonic stages and some endothelial tissues in the adult. Applications Reported: The Avas12a1 antibody has been reported for use in flow cytometric analysis. Applications Tested: The Avas12a1 antibody has been tested by flow cytometric analysis of bEnd.3 cells. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Filtration: 0.2 μm post-manufacturing filtered.

KDR (FLK1, VEGFR2) is a type III receptor tyrosine kinase. It binds vascular endothelial growth factor with high affinity in vitro, and is expressed early in development by endothelial cell precursors. FLK1 (VEGF Receptor 2) encodes one of the two receptors of the VEGF. This receptor, known as kinase insert domain receptor, is a type III receptor tyrosine kinase. The signalling and trafficking of this receptor are regulated by multiple factors, including Rab GTPase, P2Y purine nucleotide receptor, integrin alphaVbeta3, T-cell protein tyrosine phosphatase, etc. Mutations of FLK1 (VEGF Receptor 2) are implicated in infantile capillary hemangiomas.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD309 (FLK1)
Applications Flow Cytometry
Classification Monoclonal
Clone Avas12a1
Concentration 0.5 mg/mL
Conjugate Biotin
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene KDR
Gene Accession No. P35918
Gene Alias 6130401C07; CD309; CD309 antigen; fetal liver kinase 1; fetal liver kinase-1; FLK1; Flk-1; FLK1 kinase insert domain receptor (a type III receptor tyrosine kinase) (VEGF receptor 2); FLK1 kinase insert domain receptor (VEGF receptor 2); flk-1 type VEGF receptor; KDR; kinase insert domain protein receptor; kinase insert domain receptor; kinase insert domain receptor (a type III receptor tyrosine kinase); kinase NYK; Krd-1; Ly73; protein-tyrosine kinase receptor flk-1; sCD309; soluble CD309; soluble vascular endothelial growth factor receptor 2; soluble VEGFR 2; soluble VEGFR2; sVEGFR-2; tyrosine kinase growth factor receptor; vascular endotheial growth factor 2; vascular endothelial growth factor receptor 2; vascular endothelial growth factor receptor- 2; vascular endothelial growth factor receptor-2; vascular endothelial growth factor receptor-3; VEGF R2; VEGF receptor-2; VEGFR; VEGFR2; VEGFR-2
Gene Symbols KDR
Host Species Rat
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16542
Target Species Mouse
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.