Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD31 Mouse anti-Human,Baboon,Cynomolgus,Rhesus Ma, BB700, Clone: WM59 (also known as WM-59), BD Horizon™

Catalog No. p-7147131 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB566564 25 Tests
BDB566563 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB566564 Supplier BD Biosciences Supplier No. 566564
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD31 Monoclonal antibody specifically detects CD31 in Human,Baboon,Cynomolgus,Rhesus Ma samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD31
Applications Flow Cytometry
Classification Monoclonal
Clone WM59 (also known as WM-59)
Conjugate Brilliant Blue 700 (BB700)
Host Species Mouse
Quantity 25 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human, Baboon, Cynomolgus, Rhesus Macaque
Content And Storage Refrigerate (2 to 8°C)
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.