Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD31/PECAM-1 Antibody (JC/70A) - BSA Free, Novus Biologicals™
SDP

Catalog No. NB60056201M
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB60056201M 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB60056201M Supplier R&D Systems Supplier No. NB6005620.1ML

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody has been used in 34 publications

CD31/PECAM-1 Monoclonal specifically detects CD31/PECAM-1 in Feline, Human, Mouse, Rabbit, Rat samples. It is validated for Dual RNAScope ISH-IHC, ELISA, Flow Cytometry, Flow Cytometry (Cell Surface), Gene Knock-Out, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunohistochemistry Whole-Mount, Western Blot.

Specifications

Antigen CD31/PECAM-1
Applications Dual RNAScope ISH-IHC, ELISA, Flow Cytometry, Flow Cytometry (Cell Surface), Gene Knock-Out, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunohistochemistry (Whole-Mount), Western Blot
Classification Monoclonal
Clone JC/70A
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:500, Flow Cytometry 1:100-1:250, ELISA, Immunohistochemistry 1:10-1:25, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1:10-1:25, Immunohistochemistry-Frozen, Flow (Cell Surface) 1:100-1:250, Immunocytochemistry 1:25
Formulation PBS with 0.02% Sodium Azide
Gene Accession No. P16284
Gene Alias adhesion molecule, CD31, CD31 antigen, CD31/EndoCAM, EndoCAM, FLJ34100, FLJ58394, GPIIA', jc70a cd31, jc70a clone, PECA1, PECAM-1, PECAM-1, CD31/EndoCAM, platelet endothelial cell adhesion molecule, platelet/endothelial cell adhesion molecule
Gene Symbols PECAM1
Host Species Mouse
Immunogen Membrane preparation of a spleen from a patient with hairy cell leukemia.
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Cancer, Cellular Markers, Cytoskeleton Markers, Embryonic Stem Cell Markers, Endothelial Cell Markers, Extracellular Matrix, Hematopoietic Stem Cell Markers, Immunology, Mesenchymal Stem Cell Markers, Myeloid Cell Markers, Myeloid derived Suppressor Cell, Signal Transduction, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 5175
Test Specificity Human and rabbit. Rabbit data from customer review.
Target Species Feline, Human, Mouse, Rabbit, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. After opening store under sterile conditions.
Form Supernatant
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.