Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ CD31/PECAM-1 (Endothelial Cell Marker) Ab-1, Mouse Monoclonal Antibody

Catalog No. p-4545660 Shop All Epredia Products
Change view
Click to view available options
Quantity:
500 μL
1 mL
12 mL
Form:
Concentrated
Prediluted
Ready-to-Use
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form
MS353S1 500 μL Concentrated
MS353S 1 mL Concentrated
MS353RQ 12 mL Ready-to-Use
MS353R1 1 mL Prediluted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. MS353S1 Supplier Epredia™ Supplier No. MS353S1
Only null left
Add to Cart
Add to Cart

Obtain accurate, reproducible results in immunohistochemistry and immunofluorescence procedures.

CD31 is a glycoprotein expressed on endothelial cells and in platelets. It is known to be involved in cell signaling and cell adhesion.

Host Species: Mouse

Clone: JC/70A

Isotype: IgG1/κ

Species Reactivity: Human. Does not react with rat and pig. Others not tested.

Epitope: Not determined

Immunogen: Membrane preparation of a spleen from a patient with hairy cell leukemia

Molecular Weight: 100kDa (endothelial cells) and 130kDa (platelets)

Positive Control: Placenta, Tonsil

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting (Not suitable)
  • Immunofluorescence

Specifications

Antigen CD31 (Endothelial Cell Marker) Ab-1
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone JC/70A
Conjugate Unconjugated
Host Species Mouse
Immunogen Membrane preparation of a spleen from a patient with hairy cell leukemia
Quantity 500 μL
Regulatory Status IVD
Research Discipline Oncogenesis
Primary or Secondary Primary
Target Species Human
Form Concentrated
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.