Learn More
Invitrogen™ CD314 (NKG2D) Monoclonal Antibody (5C6), eBioscience™, Invitrogen™
Description
The 5C6 MAb reacts with the human NKG2D, a 42 kDa lectin-like molecule expressed by NK cells, gammadelta T cells, some CD4^+ and CD8^+ T cells. Human NKG2D forms complexes with DAP10, a membrane adaptor protein, and has the ability to costimulate multiple NK activation receptors. The counter-receptor for human NKG2D has been identified as MICA/MICB expressed on epithelial tumors from lung, breast, kidney, ovary, prostate and colon carcinoma. 5C6 and 1D11 block binding of soluble MICA to gammadelta TCR T cell clones and inhibit lysis by these cells. 5C6 and 1D11 induced NKG2D function of redirected lysis of FcReceptor bearing P815 cells.
Specifications
Specifications
| Antigen | CD314 (NKG2D) |
| Applications | Flow Cytometry, Functional Assay, Immunohistochemistry, Immunoprecipitation, Neutralization |
| Classification | Monoclonal |
| Clone | 5C6 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.2 |
| Gene | Klrk1 |
| Gene Accession No. | P26718 |
| Gene Alias | CD314; D12S2489E; D6H12S2489E; killer cell lectin like receptor K1; Killer cell lectin-like receptor subfamily K member 1; killer cell lectin-like receptor subfamily K, member 1; KLR; Klrk1; natural killer cell group 2D; natural killer cell receptor; NK cell receptor D; NK lectin-like receptor; Nkg2d; NKG2-D; NKG2-D type II integral membrane protein; NKG2-D-activating NK receptor; NKLLR; Nkrp2; NKR-P2; NKR-P2, ortholog of human NKG2D |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.