Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD315 Rat anti-Human, Clone: 17H1, MilliporeSigma™

Catalog No. MABT883MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABT883MI 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABT883MI Supplier MilliporeSigma Supplier No. MABT883
Only null left
Add to Cart
Add to Cart

Rat Monoclonal Antibody

This rat monoclonal Anti-CD315 antibody, clone 17H1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD315
Applications Flow Cytometry, Immunohistochemistry
Classification Monoclonal
Clone 17H1
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Gene Symbols PTGFRN,CD9P1,EWIF,FPRP,KIAA1436
Host Species Rat
Immunogen Mesothelioma cell-derived exosomes.
Purification Method Protein G Purified
Quantity 200 μL
Regulatory Status RUO
Research Discipline Cell Structure
Primary or Secondary Primary
Gene ID (Entrez) NP_065173.2
Test Specificity Clone 17H1 detects CD315 in A549 cells by Flow cytometry.
Target Species Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.