Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD328 (Siglec7) Monoclonal Antibody (eBioQA79 (QA79)), PE, eBioscience™
GREENER_CHOICE

Catalog No. 5010849
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-108-49 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-108-49 Supplier Invitrogen™ Supplier No. 12575942
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Description: The eBioQA79 monoclonal antibody reacts with adhesion inhibitory receptor molecule 1 (AIRM1), also known as sialic acid binding immunoglobulin-like lectin 7 (Siglec-7), p75 and CDw328. AIRM1 is a cell surface receptor that belongs to the sialoadhesin family and displays homology with the myeloid cell antigen CD33. p75/AIRM1 expression is confined to NK cells in lymphoid cells. eBioQA79 crosslinking inhibits NK function. p75/AIRM1 is expressed at various stages of myeloid differentiation. Presence of this protein is a useful way to monitor cellular proliferation and determine proliferative states of cells, especially in diseases such as chronic myeloid leukemia. Applications Reported: This eBioQA79 (QA79) antibody has been reported for use in flow cytometric analysis. Applications Tested: This eBioQA79 (QA79) antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (0.5 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 488-561 nm; Emission: μ8 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser. Filtration: 0.2 μm post-manufacturing filtered.

Putative adhesion molecule that mediates sialic-acid dependent binding to cells. Preferentially binds to alpha-2,3- and alpha-2,6-linked sialic acid. Also binds disialogangliosides (disialogalactosyl globoside, disialyl lactotetraosylceramide and disialyl GalNAc lactotetraoslylceramide). The sialic acid recognition site may be masked by cis interactions with sialic acids on the same cell surface. In the immune response, may act as an inhibitory receptor upon ligand induced tyrosine phosphorylation by recruiting cytoplasmic phosphatase(s) via their SH2 domain(s) that block signal transduction through dephosphorylation of signaling molecules. Mediates inhibition of natural killer cells cytotoxicity. May play a role in hemopoiesis. Inhibits differentiation of CD34+ cell precursors towards myelomonocytic cell lineage and proliferation of leukemic myeloid cells (in vitro).
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD328 (Siglec7)
Applications Flow Cytometry
Classification Monoclonal
Clone eBioQA79 (QA79)
Concentration 5 μL/Test
Conjugate PE
Formulation PBS with BSA and 0.09% sodium azide; pH 7.2
Gene SIGLEC7
Gene Accession No. Q9Y286
Gene Alias Adhesion inhibitory receptor molecule 1; adhesion inhibitory receptor molecule 1, siglec-7; AIRM1; AIRM-1; CD328; CDw328; D-siglec; p75; p75/ AIRM-1; p75/AIRM1; QA79; QA79 membrane protein; sialic acid binding Ig like lectin 7; sialic acid binding Ig-like lectin 7; sialic acid binding immunoglobulin-like lectin 7; sialic acid-binding Ig-like lectin 7; Siglec; SIGLEC19P; SIGLEC7; SIGLEC-7; SIGLECP2
Gene Symbols SIGLEC7
Host Species Mouse
Purification Method Affinity chromatography
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27036
Target Species Human
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.