Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD329 (Siglec-9) Mouse anti-Human, Brilliant Violet 421, Clone: E10-286, BD Optibuild™

Catalog No. BDB743363 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB743363 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB743363 Supplier BD Biosciences Supplier No. 743363
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The E10-286 monoclonal antibody specifically binds to CD329. CD329 is also known as Sialic acid-binding Ig-like lectin 9 (Siglec-9), formerly known as Siglec-Y, which is expressed on peripheral blood monocytes, granulocytes and NK cells. Siglecs (sialic acid/immunolgobulin/lectin) are a family of I-type lectins binding to sialic acids on the cell surface. They are a family of carbohydrate-binding proteins within the immunoglobulin superfamily. Siglecs are all integral membrane proteins with extracellular domains consisting of unusual N-termial V-set Ig domains, followed by variable numbers of C2-set Ig domains.

The antibody was conjugated to BD Horizon™ BV421 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 407nm and Em Max at 421nm, BD Horizon BV421 can be excited by the violet laser and detected in the standard Pacific Blue™ filter set (eg, 450/50nm filter). BD Horizon BV421 conjugates are very bright, often exhibiting a 10 fold improvement in brightness compared to Pacific Blue conjugates. BV421 is a polymer-based dye excited by the violet laser and is one of the brightest fluorochromes offered by BD Biosciences. Conjugates are typically 10 times brighter than Pacific Blue™ conjugates and are often as bright as or brighter than PE conjugates. Due to nearly identical excitation and emission properties but different spillover characteristics, BD Horizon BV421, Pacific Blue™, and BD Horizon™ V450 cannot be used simultaneously.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD329 (Siglec-9)
Classification Monoclonal
Clone E10-286
Concentration 0.2mg/mL
Conjugate Brilliant Violet 421
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias Siglec-9, Siglec Y, FOAP-9, OBBP-LIKE
Host Species Mouse
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.