Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD32b Monoclonal Antibody (AT130-2), APC, eBioscience™, Invitrogen™
GREENER_CHOICE

Catalog No. 501123043
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-112-3043 100 μg
50-112-3042 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-112-3043 Supplier Invitrogen™ Supplier No. 17032182
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

Description: The monoclonal antibody AT130-2 reacts with mouse CD32b, also known as Fc-gamma receptor IIb. CD32b is the only inhibitory member of the Fc-gamma receptor family. Immune complex engagement of CD32b results in the phosphorylation of its intracellular ITIM motif. This engagement serves to attenuate B cell receptor (BCR) signaling and other activating signals, reduces proliferation, reduces antibody production, and may also deliver apoptotic signals. CD32b is the only Fc-gamma receptor expressed by B cells. CD32b is also expressed on monocytes, dendritic cells, neutrophils, eosinophils and basophils, as well as B cell progenitors and plasma cells. AT130-2 has been reported to have agonist activity. Applications Reported: This AT130-2 antibody has been reported for use in flow cytometric analysis. Applications Tested: This AT130-2 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser. Filtration: 0.2 μm post-manufacturing filtered.

This gene encodes one member of a family of immunoglobulin Fc receptor genes found on the surface of many immune response cells. The protein encoded by this gene is a cell surface receptor found on phagocytic cells such as macrophages and neutrophils, and is involved in the process of phagocytosis and clearing of immune complexes. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD32b
Applications Flow Cytometry
Classification Monoclonal
Clone AT130-2
Concentration 0.2 mg/mL
Conjugate APC
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene FCGR2B
Gene Accession No. P08101
Gene Alias AI528646; CD32; CD32 receptor 2; CD32 receptor 2a; CD32 receptor 2b; CD32A; CD32B; CDw32; F630109E10Rik; Fc fragment of IgG receptor IIb; Fc fragment of IgG, low affinity II, receptor for (CD32); Fc fragment of IgG, low affinity IIb, receptor; Fc fragment of IgG, low affinity IIb, receptor (CD32); Fc fragment of IgG, low affinity IIb, receptor for (CD32); fc gamma receptor IIB; Fc gamma RIIb; Fc receptor, IgG, low affinity IIb; Fc[g]RII; FCG2; FC-gamma RII; fc-gamma RII-a; Fc-gamma RII-b; Fc-gamma-RII; fc-gamma-RIIa; FcgammaRIIB; fc-gamma-RIIb; FcgammaRIIB2; FcGR; Fcgr2; Fcgr2a; FCGR2A1; FCGR2B; FcgRII; Fcr-2; Fcr-3; fcRII; fcRII-a; fcRII-b; FGFR2B; IGFR2; IgG Fc receptor II beta; igG Fc receptor II-b; Low affinity immunoglobulin gamma Fc region receptor II; low affinity immunoglobulin gamma Fc region receptor II-b; Ly-17; LyM-1; Ly-m20; Lymphocyte antigen 17
Gene Symbols FCGR2B
Host Species Mouse
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14130
Target Species Mouse
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.