Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD33, Mouse anti-Human, Clone: P67.6, Millipore Sigma™

Catalog No. p-7162936 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF216325 25 μg
MABF2163100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABF216325 Supplier MilliporeSigma Supplier No. MABF216325UG
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-CD33, clone P67.6, Cat. No. MABF2163, is a mouse monoclonal antibody that detects CD33 and has been tested for use in Flow Cytometry, Immunocytochemistry, and Inhibition assays.

Myeloid cell surface antigen CD33 (UniProt: P20138; also known as Sialic acid-binding Ig-like lectin 3, Siglec-3, gp67, CD33) is encoded by the CD33 (also known as SIGLEC3) gene (Gene ID: 945) in human. CD33 is a single-pass type I membrane sialoadhesion protein that is expressed by myeloid stem cells, myeloblasts, monoblasts, monocytes, macrophages, and granulocyte precursors and preferentially binds to alpha-2,6-linked sialic acid. CD33 is not expressed in erythrocytes, platelets, B-cells, T-cells, and NK cells. Hence, it can serve as a marker for myeloid cells. It is synthesized with a signal peptide (aa 1-17), which is subsequently cleaved off in the mature form. It contains an extracellular domain (aa 18-259), a transmembrane domain (aa 260-282), and a cytoplasmic domain (aa 283-364). CD33 contains two copies of a cytoplasmic motif (aa 338-343 and 356-361), which is referred to as the immunoreceptor tyrosine-based inhibitor motif (ITIM). This motif is involved in modulation of cellular responses. In the immune response, CD33 may act as an inhibitory receptor upon ligand induced tyrosine phosphorylation by recruiting cytoplasmic phosphatase(s) via their SH2 domain(s) that block signal transduction through dephosphorylation of signaling molecules. Human CD33 is reported to contain two tyrosine residues in its cytoplasmic domain (tyrosine 340 and 358) that when phosphorylated could function as docking sites for the phosphatases, SHP-1 and/or SHP-2, and enable CD33 to function as an inhibitory receptor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD33
Applications Flow Cytometry, Immunocytochemistry, Inhibition Assays
Classification Monoclonal
Clone P67.6
Conjugate Unconjugated
Dilution Inhibition Assays: A representative lot of CD33 conjugated to calicheamicin inhibited inhibits the formation of colonies from AML marrow samples. (Hamann, P.R., et. al. (2002). Bioconjug Chem. 13(1):47-58). Flow Cytometry Analysis: A representative lot detected CD33 in Flow Cytometry applications (Kussick, S.J., et. al. (2003). Arch Pathol Lab Med. 127(9):1140-7).Immunocytochemistry Analysis: A representative lot detected CD33 in Immunocytochemistry applications (van Der Velden, V.H., et. al. (2001). Blood. 97(10):3197-204).
Formulation Purified mouse monoclonal antibody IgG1 in PBS without azide.
Gene Alias Myeloid cell surface antigen CD33;Sialic acid-binding Ig-like lectin 3;Siglec-3;gp67
Gene Symbols CD33;SIGLEC3
Host Species Mouse
Immunogen FMY9S5 cells expressing CD33.
Purification Method Protein G purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Inflammation & Immunology
Primary or Secondary Primary
Gene ID (Entrez) NP_001763.3
Test Specificity Clone P67.6 specifically detects human CD 33.
Target Species Human
Content And Storage Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.