Learn More
STEMCELL Technologies Anti-Human CD33 Antibody, Clone P67.6, PE, Size: 100 Tests
SDP
Supplier:  STEMCELL Technologies 60126PE
Encompass_Preferred
Clone P67.6 is a mouse monoclonal antibody that reacts with CD33 (Siglec-3) a 67 kDa type 1 transmembrane sialoadhesin protein of the immunoglobulin superfamily. Verified for use with flow cytometry assays use this antibody in applications requiring the detection of human CD33. CD33 is a major marker used to differentiate myeloid from lymphoid or erythroid leukemias and functions as a sialic acid-dependent cell adhesion molecule mediating protein-glycan and protein-protein interactions between cells. It also has a putative role as an inhibitory receptor during the immune response. Visit STEMCELL.com for more information and related products.
Catalog No. 50-239-0392
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.