Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD335 (NKp46) Monoclonal Antibody (9E2), PE, eBioscience™
GREENER_CHOICE

Catalog No. 12335941
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12-335-941 25 Tests
12-335-942 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 12-335-941 Supplier Invitrogen™ Supplier No. 12335941
Add to Cart
Edge
Add to Cart

Mouse Monoclonal Antibody

DescriptionThis 9E2 monoclonal antibody reacts with human CD335, which is also known as NKp46. Expressed on NK cells, CD335 is a member of the natural cytotoxicity receptor family that also includes NKp30 and NKp44. This type I transmembrane glycoprotein is involved in activating and expanding resting NK cells following interaction with dendritic cells to eliminate tumor and virus-infected (e.g., influenza) cells. CD335 also interacts with pancreatic beta cells during the development of type I diabetes. The 9E2 antibody has been reported to induce cytotoxicity.Applications Tested: This 9E2 antibody has been pre-diluted and tested by flow cytometric analysis of normal human peripheral blood cells. This may be used at 0.06 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser.

CD335, also known as NKp46 or NCR1, is a member of the natural cytotoxicity receptor family, which includes NKp30 and NKp44. This type I transmembrane glycoprotein is expressed on NK cells and plays a crucial role in activating and expanding resting NK cells through interactions with dendritic cells. CD335 enhances the ability of NK cells to eliminate tumor and virus-infected cells, such as those infected by influenza. CD335 consists of two Ig-like domains and is related to leukocyte immunoglobulin-like receptors (LIR) and killer inhibitory receptors (KIR). It contributes to the increased efficiency of activated NK cells in mediating tumor cell lysis. Additionally, CD335 interacts with pancreatic beta cells during the development of type 1 diabetes, highlighting its involvement in both immune defense and autoimmune processes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD335 (NKp46)
Applications Flow Cytometry
Classification Monoclonal
Clone 9E2
Concentration 5 μL/Test
Conjugate PE
Formulation PBS with BSA and 0.09% sodium azide; pH 7.2
Gene NCR1
Gene Accession No. O76036
Gene Alias 1-Mar; activating receptor 1; Ar1; CD335; hNKp46; Kilr1; KILR-1; LY94; Ly-94; lymphocyte antigen 94; Lymphocyte antigen 94 homolog; lymphocyte antigen 94 homolog (activating NK-receptor; lymphocyte antigen 94 homolog (activating NK-receptor; NK-p46); mAR-1; MAR1 (mouse activating receptor 1); mNKp46; natural cytotoxicity triggering receptor 1; Natural killer cell p46-related protein; natural killer cell receptor; Ncr1; NK cell-activating receptor; NK receptor KILR-1; NKACTR; Nkp46; NK-p46; NK-p46); rAR-1; rat-activating receptor 1
Gene Symbols NCR1
Host Species Mouse
Purification Method Affinity chromatography
Quantity 25 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9437
Target Species Human
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.