Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD337 (NKp30) Monoclonal Antibody (AF29-4D12), PE, eBioscience™, Invitrogen™
GREENER_CHOICE

Catalog No. 12337942
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12-337-942 100 Tests
12-337-941 25 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 12-337-942 Supplier Invitrogen™ Supplier No. 12337942
  • $366.00
    $274.65 / Each of 1

    Save $91.35

    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

Description: This AF29-4D12 monoclonal antibody reacts with human CD337, which is also known as NKp30. Expressed on NK cells, CD337 is the predominant member of the natural cytotoxicity receptor family that also includes NKp46 and NKp44. CD337 is involved in activating and expanding resting NK cells following interaction with dendritic cells to eliminate tumor, virus-infected, and immature dendritic cells. Several ligands for this receptor have been reported, including B7-H6. The AF29-4D12 antibody has been reported to crossreact to rhesus macaque. Applications Reported: This AF29-4D12 antibody has been reported for use in flow cytometric analysis. Applications Tested: This AF29-4D12 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (0.125 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser. Filtration: 0.2 μm post-manufacturing filtered.

The protein encoded by this gene is a natural cytotoxicity receptor (NCR) that may aid NK cells in the lysis of tumor cells. The encoded protein interacts with CD3-zeta (CD247), a T-cell receptor. A single nucleotide polymorphism in the 5' untranslated region of this gene has been associated with mild malaria suceptibility. Three transcript variants encoding different isoforms have been found for this gene.[provided by RefSeq, May 2010].
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD337 (NKp30)
Applications Flow Cytometry
Classification Monoclonal
Clone AF29-4D12
Concentration 5 μL/Test
Conjugate PE
Formulation PBS with BSA and 0.09% sodium azide; pH 7.2
Gene Ncr3
Gene Accession No. O14931, Q8MJ02
Gene Alias 1C7; Activating natural killer receptor p30; activating NK-A1 receptor; CD337; LY117; lymphocyte antigen 117; MALS; Natural cytotoxicity triggering receptor 3; natural killer cell p30-related protein; natural killer cell receptor; Ncr3; NK receptor 1c7; Nkp30; NK-p30; NKp30S; NKp30v1; NKp30v1ED
Gene Symbols Ncr3
Host Species Mouse
Purification Method Affinity chromatography
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 259197, 715574
Target Species Human, Monkey, Rhesus Macaque
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.