Learn More
Genetex Inc CD34 Monoclonal Antibody [MSVA-034R] HistoMAX™ (GTX04386) GeneTex - IHC-Optimized Antibody.
SDP
Supplier:  Genetex Inc GTX04386
Encompass_Preferred
The CD34 antibody [MSVA-034R] HistoMAX™ (GTX04386) is a Rabbit Monoclonal antibody in the GeneTex HistoMAX portfolio. The antibody is extensively evaluated for IHC-P using formalin-fixed human normal tissues (more than 70 different sections) and cancer tissue arrays to guarantee specificity and best-in-class performance. Information regarding both manual and autostainer staining protocols are available.Figure Legend: IHC-P analysis of human liver tissue using GTX04386 CD34 antibody [MSVA-034R] HistoMAX. CD34 is only seen in portal vessels and periportal sinusoidal.
Catalog No. NC3666323